Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A/B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | S100A/B |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
S100A/B Polyclonal specifically detects S100A/B in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
S100A/B | |
Unconjugated | |
RUO | |
P23297, P56565 | |
6271 | |
Affinity purified S-100 protein isolated from cow brain. | |
Primary | |
Store at 4°C. |
Polyclonal | |
Rabbit | |
Astrocyte Markers, Breast Cancer, Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
S100A1 | |
IgG | |
Affinity Purified | |
This antibody reacts with cow S-100 protein a and b. It shows strong crossreactivity with human S-100 a and b. In the brain, it labels glial and ependymal cells. Melanocytes and Langerhans cells of the skin stain positive with this antibody. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title