Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A1 Antibody (148), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290505F
Description
S100A1 Monoclonal specifically detects S100A1 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
S100A1 | |
Monoclonal | |
FITC | |
protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A1 (P56565; Gly2-Ser94). | |
0.1 mL | |
Breast Cancer, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction | |
6271 | |
Mouse | |
Purified |
ELISA | |
148 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Mouse S100B | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction