Learn More
S100A1 Mouse anti-Human, Clone: OTI6G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800811

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Specifications
S100A1 | |
Monoclonal | |
Unconjugated | |
S100A1 | |
S100, S100-alpha, S100A | |
Mouse | |
Affinity Chromatography | |
RUO | |
6271 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6G4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P23297 | |
S100A1 | |
Full length human recombit protein of human S100A1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.