Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01671898
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-898 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-898 Supplier Invitrogen™ Supplier No. PA110878
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

PA1-10878 detects S100 alpha-1 (Calcium Binding Protein) from bovine and porcine. PA1-10878 has been successfully used in Western blot and immunohistochemistry applications. The PA1-10878 immunogen is Alum denatured porcine S100A1.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration ∼1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene S100A1
Gene Accession No. P02639
Gene Alias AI266795; Protein S100-A1; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium-binding protein A1; S-100 protein alpha chain; S-100 protein subunit alpha; S100 protein, alpha polypeptide; S100a; S100A1; S100-alpha
Gene Symbols S100A1
Host Species Sheep
Immunogen Alum denatured porcine S100A1.
Purification Method Ammonium sulfate precipitation
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106504218, 528735
Target Species Bovine, Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.