Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S100A10 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA175390
Description
S100A10 Polyclonal Antibody for ELISA
The S-100 family of calcium-activated proteins interact with a range of target proteins to modulate biological signaling pathways. Numerous cancer cell lines overexpress the plasminogen receptor S-100A10 on the extracellular cell surface, where it forms a heterotetrameric complex with Annexin II, though this association is not required for plasma membrane localization or binding and activation of plasminogen. Additionally, S-100A10 acts as a cellular chaperone for hepatitis B (Hep B) virus polymerase. Hep B virus polymerase normally localizes to the cytoplasm only, though in the presence of S-100A10 a portion relocates to the nucleus, implying a role for S-100A10 and intracellular calcium in the process of viral replication.
Specifications
S100A10 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.2 | |
P60903 | |
S100A10 | |
Recombinant human S100A10. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
S100A10 | |
42C; AA409961; AL024248; Annexin II; annexin II ligand; annexin II ligand, calpactin I, light polypeptide; annexin II tetramer (AIIt) p11 subunit; ANX2L; ANX2LG; Ca[1]; CAL12; Cal1l; calcium binding protein A11 (calgizzarin); calpactin I light chain; Calpactin-1 light chain; cellular ligand of annexin II; CLP11; GP11; MGC111133; MGC133268; Nerve growth factor-induced protein 42C; OTTHUMP00000015270; P PRSS26; p10; p10 protein; P11; p11 subunit; Protein S100 A10; Protein S100-A10; S100 calcium binding protein A10; S100 calcium binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S100 calcium binding protein A10 (calgizzarin); S100 calcium binding protein A10 (calpactin); S100 calcium-binding protein A10; S100 calcium-binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S-100 related protein, clone 42C; S100a10 | |
Sheep | |
Affinity Chromatography | |
RUO | |
6281 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction