Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A10 Rabbit anti-Human, Clone: JF0987, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267122
Description
S100A10 Monoclonal specifically detects S100A10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
S100A10 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
annexin II ligand, calpactin I, light polypeptide, ANX2L, ANX2LG, CAL1LGP11,42C, Calpactin I light chain, Calpactin-1 light chain, Cellular ligand of annexin II, CLP11Ca[1], MGC111133, p10, p10 protein, p11, protein S100-A10, S100 calcium binding protein A10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)), S100 calcium-binding protein A10, S100 calcium-binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)) | |
Rabbit | |
Protein A purified | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
JF0987 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
S100A10 | |
Synthetic peptide within Human S100A10 aa 19-68/ 97. (SwissProt: P60903 Human; SwissProt: P08207 Mouse) | |
100 μL | |
6281 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction