Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A10 Rabbit anti-Human, Unconjugated, Clone: AODO-19, Abnova™

Catalog No. 89337087 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-087 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-087 Supplier Abnova Corporation Supplier No. MAB20687
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

S100A10 Monoclonal antibody specifically detects S100A10 in Human samples. It is validated for IHC, WB-Ce.

Specifications

Antigen S100A10
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone AODO-19
Conjugate Unconjugated
Description S100 calcium binding protein A10
Gene S100A10
Gene Alias 42C, ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, MGC111133, P11, p10
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human S100A10
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6281
Target Species Human
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.