Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A11 Antibody (118), Janelia Fluor™ 549 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289810JF549
Description
S100A11 Monoclonal antibody specifically detects S100A11 in Human samples. It is validated for ELISASpecifications
S100A11 | |
Monoclonal | |
Janelia Fluor 549 | |
calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100CS100 calcium binding protein A11 (calgizzarin) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A11 (NP_005611.1; Met1-Thr105). | |
0.1 mL | |
Cell Cycle and Replication | |
6282 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
118 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction