Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A11 Antibody (20D11), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259472PCP
This item is not returnable.
View return policy
Description
S100A11 Monoclonal antibody specifically detects S100A11 in Human samples. It is validated for Flow CytometrySpecifications
| S100A11 | |
| Monoclonal | |
| PerCP | |
| calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100CS100 calcium binding protein A11 (calgizzarin) | |
| Recombinant human S100A11 (1-105aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6282 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| 20D11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction