Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583628
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583628 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583628 Supplier Invitrogen™ Supplier No. PA583628
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLIAVFQKYA GKDGYNYTLS KTEFLSFMNT ELAAFTKNQK DPGVLDRMMK KLDTNSDGQL DFSEFLNLIG GLAMACHDSF Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 89%.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene S100A11
Gene Accession No. P31949
Gene Alias cal; Calgizzarin; calizzarin; EMAP; Emap1; EMAPI; endothelial monocyte activating polypeptide 1; endothelial monocyte-activating polypeptide; epididymis secretory protein Li 43; HEL-S-43; Metastatic lymph node gene 70 protein; MLN 70; MLN70; Protein S100-A11; Protein S100-A11, N-terminally processed; Protein S100C; Protein S100-C; S100 calcium binding protein A11; S100 calcium binding protein A11 (calgizzarin); S100 calcium binding protein A11 (calizzarin); S100 calcium binding protein A14; S100 calcium-binding protein A11; S100 calcium-binding protein A11 (calgizzarin); S100a11; S100a14; S100C
Gene Symbols S100A11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human S100A11. Recombinant protein control fragment (Product #RP-99807).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6282
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.