Learn More
Invitrogen™ S100A12 Monoclonal Antibody (161205)

Description
This antibody detects recombinant human EN-RAGE in direct ELISAs. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | S100A12 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 161205 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | S100A12 |
| Gene Accession No. | P80511 |
| Gene Alias | CAAF1; CAGC; Calcitermin; calcium-binding protein in amniotic fluid 1; calgranulin C; calgranulin-C; CGRP; EN RAGE; ENRAGE; EN-RAGE; Extracellular newly identified RAGE-binding protein; migration inhibitory factor-related protein 6; MRP6; MRP-6; Neutrophil S100 protein; p6; Protein S100 A12; protein S100-A12; S100 calcium binding protein A12; S100 calcium-binding protein A12; S100 calcium-binding protein A12 (calgranulin C); S100A12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.