Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576712 Supplier Invitrogen™ Supplier No. PA576712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

S100A12 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100A12 is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities. S100A12 is also a ligand for RAGE. Interaction with RAGE on mononuclear phagocytes, lymphocytes and endothelium triggers cellular activation. Nuclear factor kappa B, a key transcription factor for inflammatory events, is activated by S100A12/RAGE. S100A12 has also been shown to have anti-microbial properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene S100A12
Gene Accession No. P80511
Gene Alias CAAF1; CAGC; Calcitermin; calcium-binding protein in amniotic fluid 1; calgranulin C; calgranulin-C; CGRP; EN RAGE; ENRAGE; EN-RAGE; Extracellular newly identified RAGE-binding protein; migration inhibitory factor-related protein 6; MRP6; MRP-6; Neutrophil S100 protein; p6; Protein S100 A12; protein S100-A12; S100 calcium binding protein A12; S100 calcium-binding protein A12; S100 calcium-binding protein A12 (calgranulin C); S100A12
Gene Symbols S100A12
Host Species Rabbit
Immunogen Recombinant full length Human S100A12.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6283
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.