Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A13 Antibody (020), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289734MFV450
Description
S100A13 Monoclonal antibody specifically detects S100A13 in Human samples. It is validated for ELISASpecifications
S100A13 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
020 | |
ELISA | |
S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A13 (Accession#: NP_001019381.1; Met1-Lys98). | |
0.1 mL | |
Cell Cycle and Replication | |
6284 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction