Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A13 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299676APC
Description
S100A13 Polyclonal antibody specifically detects S100A13 in Human samples. It is validated for ELISASpecifications
S100A13 | |
Polyclonal | |
ELISA | |
S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 | |
Produced in rabbits immunized with purified, recombinant Human S100A13 (Accession#: NP_001019381.1; Met 1-Lys 98) | |
0.1 mL | |
Cell Cycle and Replication | |
6284 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction