Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A13 Recombinant Rabbit Monoclonal Antibody (020)
GREENER_CHOICE

Catalog No. PIMA530828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530828 Supplier Thermo Scientific Supplier No. MA530828
Add to Cart
Add to Cart

S100A13 Recombinant Rabbit Monoclonal Antibody (20)

S100A13 Recombinant Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A13
Applications ELISA
Classification Recombinant Monoclonal
Clone 020
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene S100a13
Gene Accession No. Q99584
Gene Alias Protein S100-A13; S100 calcium binding protein A13; S100 calcium-binding protein A13; S100A1; S100a13
Gene Symbols S100a13
Host Species Rabbit
Immunogen Recombinant Human S100A13 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6284
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.