Learn More
S100A14 Mouse anti-Human, Clone: OTI2G9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807603

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
S100A14 gene encodes a member of the S100 protein family which contains an EF-hand motif and binds calcium. The gene is located in a cluster of S100 genes on chromosome 1. Levels of the encoded protein have been found to be lower in cancerous tissue and associated with metastasis suggesting a tumor suppressor function.Specifications
S100A14 | |
Monoclonal | |
Unconjugated | |
S100A14 | |
BCMP84, S100A15 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57402 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2G9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9HCY8 | |
S100A14 | |
Full length human recombit protein of human S100A14 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.