Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A16 Antibody (140), DyLight 488 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289804G
Description
S100A16 Monoclonal antibody specifically detects S100A16 in Human samples. It is validated for ELISASpecifications
S100A16 | |
Monoclonal | |
DyLight 488 | |
Aging-associated gene 13 protein, aging-associated protein 13, DT1P1A7, MGC17528, protein S100-A16, Protein S100-F, S100 calcium binding protein A16, S100FS100 calcium-binding protein A16 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A16 (Q96FQ6; Met1-Ser103). | |
0.1 mL | |
Cell Cycle and Replication | |
140576 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
140 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction