Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547840
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547840 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547840 Supplier Invitrogen™ Supplier No. PA547840
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 30% cross-reactivity with recombinant human S100A16 is observed, and less than 1% cross-reactivity with recombinant mouse S100A4, recombinant human (rh) S100A2, and rhS100A10 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

S100A16 (S100 Calcium Binding Protein A16) is a Protein Coding gene. Diseases associated with S100A16 include Robinow Syndrome, Autosomal Recessive and Robinow Syndrome. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity. Calcium-binding protein. Binds one calcium ion per monomer. Can promote differentiation of adipocytes (in vitro). Overexpression in preadipocytes increases their proliferation, enhances adipogenesis and reduces insulin-stimulated glucose uptake.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A16
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene S100a16
Gene Accession No. Q9D708
Gene Alias 2300002L21Rik; AAG13; aging-associated gene 13 protein; aging-associated protein 13; AI325039; AI663996; DT1P1A7; MGC17528; protein S100-A16; Protein S100F; Protein S100-F; S100 calcium binding protein A16; S100 calcium-binding protein A16; S100A16; S100F
Gene Symbols S100a16
Host Species Sheep
Immunogen E. coli-derived recombinant mouse S100A16 Ala2-Ser124.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67860
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.