Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A16 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570955
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0955 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0955 Supplier Bethyl Laboratories, Inc Supplier No. A305655AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Calcium-binding protein. Binds one calcium ion per monomer. Marenholz I., Submitted (OCT-2003) to the EMBL/GenBank/DDBJ databases. Morita K.; Submitted (JAN-2003) to the EMBL/GenBank/DDBJ databases. Gregory S.G., Nature 441:315-321(2006).

Specifications

Antigen S100A16
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene S100A16
Gene Accession No. Q96FQ6
Gene Alias S100F
Gene Symbols S100A16
Host Species Rabbit
Immunogen Between 53 and 103
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140576
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.