Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A2, Mouse anti-Human, Clone: CPTC-S100A2-2, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $526.00
Specifications
Antigen | S100A2 |
---|---|
Clone | CPTC-S100A2-2 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
S100A2 Monoclonal specifically detects S100A2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
S100A2 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cellular Markers, Neuroscience | |
10 mM PBS with 0.05% BSA | |
CAN19protein S100-A2, MGC111539, S100 calcium binding protein A2, S100 calcium-binding protein A2Protein S-100L, S100L | |
S100A2 | |
IgG2b κ | |
Protein A or G purified |
CPTC-S100A2-2 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P29034, P29034 | |
6273 | |
Recombinant full-length human S100A2 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title