Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    S100A3 Antibody (05), mFluor Violet 450 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306623MFV450
Description
S100A3 Monoclonal antibody specifically detects S100A3 in Human samples. It is validated for ELISASpecifications
| S100A3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| ELISA | |
| 05 | |
| ELISA | |
| Protein S-100E, S100 calcium binding protein A3, S100 calcium-binding protein A3S100Eprotein S100-A3 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human S100A3 (Uniprot#: P33764; Met 1-Gln 101). | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 6274 | |
| Store at 4C in the dark. | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            