Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298279JF646
Description
S100A3 Polyclonal antibody specifically detects S100A3 in Mouse samples. It is validated for Western Blot, ELISASpecifications
S100A3 | |
Polyclonal | |
Western Blot, ELISA | |
Protein S-100E, S100 calcium binding protein A3, S100 calcium-binding protein A3S100Eprotein S100-A3 | |
Produced in rabbits immunized with purified, recombinant Mouse S100A3 (Accession#: NP_035440.1; Met1-Gln101) | |
0.1 mL | |
Neuroscience, Signal Transduction | |
6274 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction