Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566001 Supplier Invitrogen™ Supplier No. PA566001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MARPLEQAVAA IVCTFQEYAG RCGDKYKLCQ AELKELLQK Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein has the highest content of cysteines of all S100 proteins, has a high affinity for Zinc, and is highly expressed in human hair cuticle. The precise function of this protein is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene S100A3
Gene Accession No. P33764
Gene Alias Protein S100-A3; Protein S-100E; S100 calcium binding protein A3; S100 calcium-binding protein A3; S100A3; S100E
Gene Symbols S100A3
Host Species Rabbit
Immunogen Recombinant Human S100A3. Recombinant protein control fragment (Product #RP-106689).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6274
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.