Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A4 Monoclonal Antibody (CL0239)
GREENER_CHOICE

Catalog No. PIMA531332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531332 Supplier Invitrogen™ Supplier No. MA531332
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: MACPLEKALD VMVSTFHKYS GKEGDKFKLN KSELKELLTR ELPSFLGKRT DEAAFQKLMS NLDSNRDNEV DFQEYCVFLS CIAMMCNEFF EGFPDKQPRK K Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 91%.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL0239
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene S100a4
Gene Accession No. P26447
Gene Alias 18A2; 42a; calvasculin; CAPL; fibroblast-specific protein-1; FSP1; leukemia multidrug resistance associated protein; malignant transformation suppression 1; metastasin; Metastatic cell protein; Mts1; Nerve growth factor-induced protein 42A; P9K; P9KA; PeL98; pk9a; Placental calcium-binding protein; pr; protein 18A2; Protein 9 Ka homologous to calcium-binding protein; protein Mts1; Protein S100-A4; RNP9KA; S100 calcium binding protein A4; S100 calcium-binding protein A4; S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog); S100a4
Gene Symbols S100a4
Host Species Mouse
Immunogen Recombinant protein fragment of Human S100A4 (Product #RP-88717)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6275
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.