Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A4 Mouse anti-Human, Mouse, Clone: 2G11B4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | S100A4 |
---|---|
Clone | 2G11B4 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6912
![]() ![]() |
Proteintech Group Inc
664891IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Specifications
S100A4 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
18A2, 42A, Calvasculin, CAPL, Fibroblast-specific protein 1, FSP1, FSP-1, Metastasin, MTS1, P9KA, PEL98, Protein Mts1, Protein S100 A4, S100A4 | |
S100A4 | |
IgG2a | |
Protein A | |
Antibody |
2G11B4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P07091, P26447 | |
20198, 6275 | |
S100A4 Fusion Protein ag9019 | |
Primary | |
-20°C | |
S100A4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title