Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $404.50
Specifications
Antigen | S100A4 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145246
|
Invitrogen™
PA5145246 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145245
|
Invitrogen™
PA5145245 |
200 μL |
Each for $404.50
|
|
|||||
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
S100A4 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P07091, P26447 | |
20198, 6275 | |
Recombinant Human S100A4 protein, Ala2-Lys101 (Accession #P26447). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
18A2; 42a; calvasculin; CAPL; fibroblast-specific protein-1; FSP1; leukemia multidrug resistance associated protein; malignant transformation suppression 1; metastasin; Metastatic cell protein; Mts1; Nerve growth factor-induced protein 42A; P9K; P9KA; PeL98; pk9a; Placental calcium-binding protein; pr; protein 18A2; Protein 9 Ka homologous to calcium-binding protein; protein Mts1; Protein S100-A4; RNP9KA; S100 calcium binding protein A4; S100 calcium-binding protein A4; S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog); S100a4 | |
S100a4 | |
IgG | |
Protein A | |
S100a4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title