Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A4 Rabbit anti-Human, Mouse, Unconjugated, Clone: S100A4/2750R, Abnova™

Catalog No. 89339601 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-601 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-601 Supplier Abnova Corporation Supplier No. RAB00622
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

S100A4 Monoclonal antibody specifically detects S100A4 in Human, Mouse samples. It is validated for WB-Ce, IHC-P, IF, Flow Cyt.

Specifications

Antigen S100A4
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone S100A4/2750R
Conjugate Unconjugated
Description S100 calcium binding protein A4
Gene S100A4
Gene Alias 18A2, 42A, CAPL, FSP1, MTS1, P9KA, PEL98
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-200 of human S100A4
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6275
Target Species Human, Mouse
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.