Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A5 Antibody (011), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290582MFV610
Description
S100A5 Monoclonal antibody specifically detects S100A5 in Mouse samples. It is validated for ELISASpecifications
S100A5 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
011 | |
ELISA | |
Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
Rabbit immunized with purified, recombinant Mouse S100A5 (Uniprot#: P63084, Met1-Lys93) | |
0.1 mL | |
Neuroscience, Signal Transduction | |
6276 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction