Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A5 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299722AF700
Description
S100A5 Polyclonal specifically detects S100A5 in Human samples. It is validated for ELISA.Specifications
S100A5 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
6276 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 700 | |
Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
Produced in rabbits immunized with purified, recombinant S100A5 (B48219; Met 1-Lys 110). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction