Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A5 Antibody (OTI4C9), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273968AF488
Description
S100A5 Monoclonal specifically detects S100A5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| S100A5 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| S100A5 | |
| Full length recombinant protein of human S100A5 (NP_002953) produced in 293T cell. | |
| 0.1 mL | |
| 6276.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI4C9 | |
| Western Blot | |
| Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction