Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A5 Monoclonal Antibody (C8), Invitrogen™

Catalog No. PIMA543596
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543596 20 μL
PIMA543595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543596 Supplier Invitrogen™ Supplier No. MA543596
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Binds calcium, zinc and copper. One subunit can simultaneously bind 2 calcium ions or 2 copper ions plus 1 zinc ion. Calcium and copper ions compete for the same binding sites. Engelkamp D., Proc. Natl. Acad. Sci. U.S.A. 90:6547-6551(1993). Gregory S.G., Nature 441:315-321(2006).
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene S100A5
Gene Accession No. P33763
Gene Alias Protein S100-A5; Protein S-100D; S100 calcium binding protein A5; S100 calcium-binding protein A4; S100 calcium-binding protein A5; S100a5; S100D; S100D9
Gene Symbols S100A5
Host Species Mouse
Immunogen Met1-Lys92 (Accession P33763), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6276
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.