Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A6 Antibody (016), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290488MFV500
Description
S100A6 Monoclonal antibody specifically detects S100A6 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISASpecifications
S100A6 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
016 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Capture | |
2A9, CABP, CACY5B10, calcyclin, Growth factor-inducible protein 2A9, MLN 4, PRAS100 calcium binding protein A6 (calcyclin), Prolactin receptor-associated protein, protein S100-A6, S100 calcium binding protein A6, S100 calcium-binding protein A6, S100 calcium-binding protein A6 (calcyclin) | |
Rabbit immunized with purified, recombinant Mouse S100A6 (Met 1-Lys 89, Accession#: NP_035443.1) | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research | |
6277 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction