Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.16 - $494.86
Specifications
| Antigen | S100A6 |
|---|---|
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11093274T
![]() |
Novus Biologicals
NB11093274SS |
0.025 mL |
Each for $182.16
|
|
|||||
NB11093274
![]() |
Novus Biologicals
NB11093274 |
0.1 mL |
Each for $494.86
|
|
|||||
Description
S100A6 Polyclonal specifically detects S100A6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| S100A6 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research | |
| Whole antisera with 0.1% Sodium Azide | |
| 2A9, CABP, CACY5B10, calcyclin, Growth factor-inducible protein 2A9, MLN 4, PRAS100 calcium binding protein A6 (calcyclin), Prolactin receptor-associated protein, protein S100-A6, S100 calcium binding protein A6, S100 calcium-binding protein A6, S100 calcium-binding protein A6 (calcyclin) | |
| S100A6 | |
| IgG | |
| Unpurified | |
| 9 kDa |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| P06703 | |
| 6277 | |
| Full length human S100A6 protein [Swiss-Prot# P06703] expressed in E. coli. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title