Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531862 Supplier Invitrogen™ Supplier No. PA531862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (96%), Rat (95%), Pig (87%), Rabbit (90%), Chicken (88%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene S100A6
Gene Accession No. P06703
Gene Alias 2A9; 5B10; CABP; Cacy; calcium binding protein A6 (calcyclin); calcyclin; Growth factor-inducible protein 2A9; lung 10 kDa protein; MLN 4; PRA; Prolactin receptor-associated protein; protein S100-A6; S100 calcium binding protein A6; S100 calcium binding protein A6 (calcyclin); S100 calcium-binding protein A6; S100 calcium-binding protein A6 (calcyclin); S100a6
Gene Symbols S100A6
Host Species Rabbit
Immunogen Full length human S100A6 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.