Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A6 Polyclonal Antibody

Catalog No. PIPA5121631
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121631 20 μL
PIPA5119404 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121631 Supplier Invitrogen™ Supplier No. PA5121631
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

S100A6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene S100A6
Gene Accession No. P06703
Gene Alias 2A9; 5B10; CABP; Cacy; calcium binding protein A6 (calcyclin); calcyclin; Growth factor-inducible protein 2A9; lung 10 kDa protein; MLN 4; PRA; Prolactin receptor-associated protein; protein S100-A6; S100 calcium binding protein A6; S100 calcium binding protein A6 (calcyclin); S100 calcium-binding protein A6; S100 calcium-binding protein A6 (calcyclin); S100a6
Gene Symbols S100A6
Host Species Rabbit
Immunogen Recombinant protein S100 Calcium Binding Protein A6. The antigen corresponds to amino acid range 1-90 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.