Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S100A7 Monoclonal Antibody (9F3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549102
Description
S100A7 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Functional Assay, IA
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus. This protein is markedly over-expressed in the skin lesions of psoriatic patients, but is excluded as a candidate gene for familial psoriasis susceptibility. The exact function of this protein is not known.
Specifications
S100A7 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and no preservative | |
P31151 | |
S100a15a, S100A7 | |
Full length recombinant human S100A7. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Functional Assay, Inhibition Assays, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
9F3 | |
Unconjugated | |
S100A7 | |
AY465109; Gm1020; NICE-2; protein S100-A15A; Protein S100-A7; protein S100-A7A; PSOR1; Psoriasin; psoriasin 1; S100 calcium binding protein A15; S100 calcium binding protein A7; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A15A; S100 calcium-binding protein A7; S100 calcium-binding protein A7 (psoriasin 1); S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100a15a; S100a17l1; S100A7; S100a7a; S100A7c; S100A7f; S100A7L1 | |
Mouse | |
Protein G | |
RUO | |
6278 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction