Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144358 Supplier Invitrogen™ Supplier No. PA5144358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S100A7 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus. This protein is markedly over-expressed in the skin lesions of psoriatic patients, but is excluded as a candidate gene for familial psoriasis susceptibility. The exact function of this protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A7
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene S100A7
Gene Accession No. P31151
Gene Alias AY465109; Gm1020; NICE-2; protein S100-A15A; Protein S100-A7; protein S100-A7A; PSOR1; Psoriasin; psoriasin 1; S100 calcium binding protein A15; S100 calcium binding protein A7; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A15A; S100 calcium-binding protein A7; S100 calcium-binding protein A7 (psoriasin 1); S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100a15a; S100a17l1; S100A7; S100a7a; S100A7c; S100A7f; S100A7L1
Gene Symbols S100a15a, S100A7
Host Species Rabbit
Immunogen Recombinant Human Protein S100-A7 protein (2-101AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6278
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.