Learn More
Invitrogen™ S100A7 Polyclonal Antibody

Description
Immunogen sequence: EKPSLLTMMK ENFPNFLSAC DKKGTNYLAD VFEKKDKNED KKIDFSEFLS LLGDIATDYH KQSHGAAPCS Highest antigen sequence indentity to the following orthologs: Mouse - 30%, Rat - 30%.
Specifications
Specifications
| Antigen | S100A7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | S100A7 |
| Gene Accession No. | P31151 |
| Gene Alias | AY465109; Gm1020; NICE-2; protein S100-A15A; Protein S100-A7; protein S100-A7A; PSOR1; Psoriasin; psoriasin 1; S100 calcium binding protein A15; S100 calcium binding protein A7; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A15A; S100 calcium-binding protein A7; S100 calcium-binding protein A7 (psoriasin 1); S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100a15a; S100a17l1; S100A7; S100a7a; S100A7c; S100A7f; S100A7L1 |
| Gene Symbols | S100a15a, S100A7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.