Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S100A7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145457
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus. This protein is markedly over-expressed in the skin lesions of psoriatic patients, but is excluded as a candidate gene for familial psoriasis susceptibility. The exact function of this protein is not known.
Specifications
S100A7 | |
Polyclonal | |
Unconjugated | |
S100A7 | |
AY465109; Gm1020; NICE-2; protein S100-A15A; Protein S100-A7; protein S100-A7A; PSOR1; Psoriasin; psoriasin 1; S100 calcium binding protein A15; S100 calcium binding protein A7; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A15A; S100 calcium-binding protein A7; S100 calcium-binding protein A7 (psoriasin 1); S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100a15a; S100a17l1; S100A7; S100a7a; S100A7c; S100A7f; S100A7L1 | |
Rabbit | |
Protein A | |
RUO | |
282344 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Q28050 | |
S100A7 | |
Recombinant Bovine S100A7 protein, Met1-Gln101 (Accession #Q28050). | |
200 μL | |
Primary | |
Bovine | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction