Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A7/Psoriasin Antibody (129), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289814JF646
Description
S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
S100A7/Psoriasin | |
Monoclonal | |
Janelia Fluor 646 | |
HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A7/Psoriasin (NP_002954.2; Met 1-Gln 101). | |
0.1 mL | |
Cancer | |
6278 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
129 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction