Learn More
Invitrogen™ S100A7 Recombinant Rabbit Monoclonal Antibody (129)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529541
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human S100A7.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus. This protein is markedly over-expressed in the skin lesions of psoriatic patients, but is excluded as a candidate gene for familial psoriasis susceptibility. The exact function of this protein is not known.
Specifications
S100A7 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P31151 | |
S100a15a, S100A7 | |
Recombinant Human S100A7 protein (Met1-Gln101). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
129 | |
Unconjugated | |
S100A7 | |
AY465109; Gm1020; NICE-2; protein S100-A15A; Protein S100-A7; protein S100-A7A; PSOR1; Psoriasin; psoriasin 1; S100 calcium binding protein A15; S100 calcium binding protein A7; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A15A; S100 calcium-binding protein A7; S100 calcium-binding protein A7 (psoriasin 1); S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100a15a; S100a17l1; S100A7; S100a7a; S100A7c; S100A7f; S100A7L1 | |
Rabbit | |
Protein A | |
RUO | |
6278 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.