Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568288 Supplier Invitrogen™ Supplier No. PA568288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S100A7A Polyclonal Antibody for Western Blot

May be involved in epidermal differentiation and inflammation and might therefore be important for the pathogenesis of psoriasis and other diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A7A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene S100A7A
Gene Accession No. Q86SG5
Gene Alias NICE-2; Protein S100-A7A; S100 calcium binding protein A15; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100A7A; S100A7f; S100A7L1
Gene Symbols S100A7A
Host Species Rabbit
Immunogen Synthesized peptide derived from human S100A7A.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338324
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.