Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A8/A9 Antibody (SPM281), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311540PE
Description
S100A8/A9 Monoclonal antibody specifically detects S100A8/A9 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline, Guinea Pig, Goat, Baboon, Monkey, Rabbit samples. It is validated for Flow CytometrySpecifications
S100A8/A9 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline, Guinea Pig, Goat, Baboon, Monkey, Rabbit | |
Purified |
Flow Cytometry | |
SPM281 | |
Flow Cytometry | |
60B8AG, CAGA, Calprotectin, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8, S100 calcium binding protein A8, S100A8 | |
Affinity Purified monocyte membrane preparation | |
0.1 mL | |
Cancer | |
6279 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction