Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8/A9 Monoclonal Antibody (OTI5A9), TrueMAB™, OriGene
SDP

Catalog No. 502543348
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3348 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3348 Supplier OriGene Supplier No. CF814395
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A8/A9 Monoclonal antibody specifically detects S100A8/A9 in Human samples. It is validated for ELISA

Specifications

Antigen S100A8/A9
Applications ELISA
Classification Monoclonal
Clone OTI5A9
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene LCN2
Gene Accession No. P80188
Gene Alias 60B8Ag; AI323541; AW546964; B8Ag; BEE11; BEE22; CAGA; CAGB; calcium binding protein; S100 protein; calgranulin A; calgranulin B; calgranulin-A; Calgranulin-B; calprotectin; calprotectin L1H subunit; Calprotectin L1L subunit; CFAG; CGLA; CGLB; chemotactic cytokine CP-10; chemotactic S100 protein; CP-10; Cystic fibrosis antigen; EGK_01322; EGK_21438; GAGB; intracellular calcium-binding protein (MRP14); L1AG; Leukocyte L1 complex heavy chain; leukocyte L1 complex light chain; LIAG; LOC100735514; LOC532569 protein; MA387; MAC387; macrophage migration inhibitory factor-related protein-14; macrophage migration inhibitory factor-related protein-8; MIF; migration inhibitory factor-related protein 14; Migration inhibitory factor-related protein 8; Mrp14; MRP-14; MRP8; MRP-8; myeloid-related protein 14; Neutrophil cytosolic 23 kDa protein; neutrophil cytosolic 7 kDa protein; NIF; p14; p23; P7; p8; Pro-inflammatory S100 cytokine; protein S100-A8; Protein S100-A9; S100 calcium binding protein A8; S100 calcium binding protein A8 (calgranulin A); S100 calcium binding protein A9; S100 calcium binding protein A9 (calgranulin B); S100 calcium binding protein A9; protein S100-A9; S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100a8; S100A8/S100A9; S100a9; similar to Calgranulin A (Migration inhibitory factor-related protein 8) (MRP-8) (Cystic fibrosis antigen) (CFAG) (P8) (Leukocyte L1 complex light chain) (S100 calcium-binding protein A8) (Calprotectin L1L subunit) (Urinary stone protein band A)...; urinary stone protein band A
Host Species Mouse
Immunogen Full length human recombinant protein of human NGAL (NP_005555) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3934
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.