Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8/A9 (MRP-8/14) Mouse anti-Human, Alexa Fluor 647, Clone: 5.5, BD Pharmingen™

Catalog No. BDB566010 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566010 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566010 Supplier BD Biosciences Supplier No. 566010
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A8/A9 (MRP-8/14) Monoclonal antibody specifically detects S100A8/A9 (MRP-8/14) in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A8/A9 (MRP-8/14)
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 5.5
Conjugate Alexa Fluor 647
Description Vol. Per test: 5μL
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias S100A8/S100A9; MRP-8/MRP-14; Calprotectin, CFAG; p8,14
Host Species Mouse
Immunogen Acute Monocytic Leukemia Cells and Monocytes
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.