Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A8 Antibody (008), Alexa Fluor™ 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290487AF594
Description
S100A8 Monoclonal specifically detects S100A8 in Mouse samples. It is validated for ELISA.Specifications
S100A8 | |
Monoclonal | |
Alexa Fluor 594 | |
60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A8 (P27005; Pro2-Glu89). | |
0.1 mL | |
Cancer | |
6279 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
8 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction