Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A8 Antibody (2C5/4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP276650B
Description
S100A8 Monoclonal specifically detects S100A8 in Human samples. It is validated for Western Blot, ELISA.Specifications
S100A8 | |
Monoclonal | |
Biotin | |
PBS with 0.05% sodium azide | |
S100A8 | |
Recombinant human S100A8 | |
0.1 mL | |
Primary | |
This antibody is specific for S100A8 both as an isolated protein and in combination with S100A9. | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
2C5/4 | |
Western Blot, ELISA | |
60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
Mouse | |
Protein A purified | |
Cancer | |
6279.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction