Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Antibody (63N13G5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP227067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227067 0.1 mg
NBP227067SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227067 Supplier Novus Biologicals Supplier No. NBP227067
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 1 publication

S100A8 Monoclonal specifically detects S100A8 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen S100A8
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 63N13G5
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry reported by customer review, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. P27005
Gene Alias 60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
Gene Symbols S100A8
Host Species Rat
Immunogen Within the range of amino acids 2-98 of mouse S100A8 were used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6279
Test Specificity The amino acid sequence used as immunogen for this antibody has an 81% homology with Rat S100A8.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.