Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human S100A8 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB4570SP
This item is not returnable.
View return policy
Description
S100A8 Monoclonal specifically detects S100A8 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
S100A8 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P05109 | |
S100A8 | |
E. coli-derived recombinant human S100A8 Met1-Glu93 Accession # P05109 | |
25 μg | |
Primary | |
Detects human S100A8 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) S100A8/A9 is observed and no cross-reactivity with rhS100A9, recombinant mouse (rm) S100A8, rmS100A9, or rmS100A8/A9 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
749916 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
60B8AG, CAGACP-10, calgranulin A, Calgranulin-A, Calprotectin L1L subunit, CFAG, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6279 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction