Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Rabbit anti-Human, Unconjugated, Clone: MAC3157R, Abnova™

Catalog No. 89339602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-602 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-602 Supplier Abnova Corporation Supplier No. RAB00623
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

S100A8 Monoclonal antibody specifically detects S100A8 in Baboon, Bovine, Cat, Dog, Goat, Guinea pig, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat samples. It is validated for IHC-P.

Specifications

Antigen S100A8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MAC3157R
Conjugate Unconjugated
Description S100 calcium binding protein A8
Gene S100A8
Gene Alias 60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8
Host Species Rabbit
Immunogen Affinity Purified monocyte membrane preparation
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6279, 6280
Target Species Baboon, Bovine, Feline, Dog, Goat, Guinea Pig, Equine, Human, Monkey, Mouse, Pig, Rabbit, Rat
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.